Microtubes without Cap, Sarstedt
Supplier: Sarstedt
72.708 72.696
101453-235PK 48.74 USD
101453-235 101093-233
Microtubes without Cap, Sarstedt
Tubes Microtubes
Transparent polypropylene microtubes are supplied without caps.
Microtubes can withstand centrifugation up to 20000 ×g.