Circulator Adapters, Ace Glass Incorporated
Supplier: Ace Glass
12187-82 12187-83 12187-58 12187-61 12187-62 12187-63 12187-70 12187-71 12187-72 12187-73 12187-74 12187-81 12187-84
75877-610EA 860.22 USD
75877-610 75877-612 75877-686 75877-688 75877-690 75877-692 75877-694 75877-696 75877-698 75877-700 75877-702 75877-704 76107-048
Circulator Adapters, Ace Glass Incorporated
Fittings and Connectors
Stainless steel beaded pipe adapters designed for use with various manufacturers' circulators, and Ace Glass reaction systems.
  • Made of 304 stainless steel
  • Adapters with hose barbs connect to hoses on a circulator
  • Adapters with "M" style fittings connect to NPT threads