N-(4-Isothiocyanatobenzyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide ≥98%

Supplier: 0000027161
Warning

CDX-I0015-M025 CDX-I0015-M050 CDX-I0015-M250
102990-154EA 127.38 USD
102990-154 102990-156 102990-158
N-(4-Isothiocyanatobenzyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide ≥98%
N-(4-Isothiocyanatobenzyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide

New class of highly photostable, water soluble fluorescent labels with large Stoke's shift, high QY in aqueous media and pH tolerance. Major disadvantage: rel. low extinction coefficient.

Formula: C₂₃H₁₈BrN₃O₂S
Storage Temperature: Freezer
CAS Number: 155862-90-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Orange powder.
Solubility Soluble in DMF or acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR