Boromycin ≥99%

Supplier: 0000027161
Warning

AG-CN2-0166-C250 AG-CN2-0166-M001
102988-122EA 166.15 USD
102988-122 102988-124
Boromycin ≥99%
Boromycin

Boron-containing macrolide antibiotic


Source/Host: Isolated from Streptomyces sp.
Purity: >99% (TLC)
Appearance/Color: Off-white solid.
Solubility: Soluble in DMSO, ethanol or methanol.


Antibacterial. Antiprotozoal (Plasmodium, Babesia). Anti-HIV compound. Ca2+ homeostasis modulator. Potentiator of bleomycin-induced anti-tumor activity. Used as a chiral selector in capillary electrophoresis.

Storage Temperature: Freezer
CAS Number: 34524-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR