Tubing Connectors, Ace Glass Incorporated
Supplier: Ace Glass
5855-08 5855-10 5855-71 5855-73 5855-80
75877-578PK 186.13 USD
75877-578 75877-580 75877-582 75877-584 75877-586
Tubing Connectors, Ace Glass Incorporated
Fittings and Connectors
Small tubing connector for use with Chromatography to tubing adapters
  • Separate PTFE nut, Kel-F wedge insert, and wedge insertion tool make up the complete connector
  • Connector components for either 1.5 mm or 2 mm I.D. tubing
  • Wedge insertion tool mechanically wedges the tubing between the insert and the inside wall of the nut
Small tubing connector comprised of a virgin TFE 1/4"–28 UNF 2B male nut and a Kel-F wedge insert. Use with ACE chromatograpy column to tubing filter adapters.
Suitable for vacuum or pressure applications, as tubing won't slip off or blow out.
Packaging: The PTFE nuts and Wedge Inserts are sold in packs of 12. The Wedge Insertion Tool is sold individually.