Rotary Diamond Micro-Engraver Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50465-15
103302-470EA 38.79 USD
103302-470
Rotary Diamond Micro-Engraver Pen, Electron Microscopy Sciences
Pens Stylus and Scribers
This micro-engraver pen may be used to mark samples, engrave tools, indicate a sample area for finding in the SEM and deburring.
  • Easy to handle and control, this micro engraver is ideal for detailed engraving on almost any surface like wood, metal, ceramic and glass
  • Contoured grip for comfort and control
  • Pen-type design fits in a shirt pocket or purse and goes anywhere
  • Precision-ground stainless steel bit with diamond point tip
  • Requires two AAA batteries
The pen measures 61/4" and weighs 0.8 kg.
Delivery information: Two AAA batteries are included.