Spunbonded Polypropylene Shoe Covers, SCR200 Series, CT International
Supplier: CT International
SCR200XL-NS SCR200XL
33502-674CS 115.46 USD
33502-674 56613-804
Spunbonded Polypropylene Shoe Covers, SCR200 Series, CT International
Disposable Garments
SCR200 Series spunbonded polypropylene shoe covers are manufactured from a lightweight material that offers breathability and comfort, while maintaining full coverage
  • Strong and durable
  • Contamination control
  • No latex proteins
  • Lot traceability
All items are individually inspected for stitching, sizing, material, quality, and workmanship.
Packaging: Stacked and rolled, 50 Pairs/Bag, 4 Bags/Case, 200 Pair/Case.