Sodium acetate trihydrate USP, Ph. Eur., BP endotoxin tested
Supplier: Avantor Performance Materials
Synonyms:
Acetic acid sodium salt trihydrate
VWRB530-12KG
VWRB530-500G
VWRB530-12KGEA
22491.35
USD
VWRB530-12KG
VWRB530-500G
Sodium acetate trihydrate USP, Ph. Eur., BP endotoxin tested
Sodium acetate trihydrate
Tested according to the USP, EP, and BP monographs. Material is of non-animal, non-human origin.
- Bioprocessing
- Suitable for use as an Excipient
Formula:
H₃CCOONa·3H₂O MW: 136.08 g/mol Boiling Pt: 120…123 °C (1013 hPa) Melting Pt: 58 Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 Merck Index: 13,08642 |
|
Specification Test Results
Assay | 99.0-101.0% |
Appearance of Solution | Pass |
Bacterial Endotoxins | ≤ 2.5 EU/g |
Calcium and Magnesium Test | Pass, ≤ 50 ppm |
Chloride |
≤ 350 ppm |
Chlorides (2.4.4) | ≤ 200 ppm |
Arsenic (2.4.2, Method A) | ≤ 2 ppm |
Heavy Metals |
≤ 10 ppm |
Identification A | Pass |
Identification B | Pass |
Identification C | Pass |
Insoluble Matter | ≤ 0.05% |
Iron (2.4.9) | ≤ 10 ppm |
Loss on Drying |
38.0 – 41.0% |
Loss on Drying (2.2.32) | 39.0 – 40.5% |
pH |
7.5 – 9.2 |
pH (2.2.3) | 7.5 – 9.0 |
Potassium Test | Pass |
Sulfate |
≤ 50ppm |
Sulfates (2.4.13) | ≤ 200ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...