D-(+)-Sucrose Ph. Eur., NF, BP, JP endotoxin tested

Supplier: Avantor Performance Materials

Synonyms: alpha-D-Glucopyranoside, β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, beta-D-Fructofuranosyl alpha-D-glucopyranoside, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, D-(+)-Saccharose, Sucrose, Sugar

VWRB335-12KG VWRB335-1KG
VWRB335-100GEA 90.63 USD
VWRB335-100G VWRB335-12KG VWRB335-1KG
D-(+)-Sucrose Ph. Eur., NF, BP, JP endotoxin tested
D(+)-Sucrose

Tested according to the USP, EP, BP and JP monographs. Material is of non-animal, non-human origin.


  • Bioprocessing
  • Suitable for use as an Excipient

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance of Solution Pass
Bacterial Endotoxins ≤ 50 EU/g
Color Value (NF) ≤ 75
Color Value (EP/BP/JP) ≤ 45
Conductivity ≤ 35 μS/cm
Dextrins Pass
Identification Pass
Loss on Drying ≤ 0.1%
Reducing Sugars Test Pass
Specific Optical Rotation (2.2.7) +66.3 - +67.0 Degrees
Sulfites ≤ 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR