Zinc stearate, powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Stearic acid zinc salt, Zinc(II) stearate

ZI115-2.5KG ZI115-12KG ZI115-500GM
95036-654EA 450.9 USD
95036-654 700005-058 95036-656 75812-644
Zinc stearate, powder USP
Zinc stearate

ZINC STEARATE, POWDER, USP is also referred to as 'Zinc Soap' and can be used in powders and ointments in the treatment of eczema, acne, and other skin diseases. It may also be used as a lubricant in cosmetic formulations to improve texture and smoothness. Zinc stearate has antiseptic, astringent and topical protective properties. Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₃₆H₇₀O₄Zn
MW: 632.34 g/mol
Melting Pt: 140
Flash Pt: 276
Storage Temperature: Ambient
MDL Number: MFCD00013031
CAS Number: 557-05-1
Merck Index: 13,10212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8ad7f3e101681798edbd54e57ad5d2ac82b2e5d&vid=435308cdcb35862fe6cc06384da2ef10d84c5700&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (ZnO) 12.5 - 14.0 %
Alkalies and Alkaline Earths max. 1.0 %
Arsenic max. 1.5 ppm
Lead max. 10 ppm
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR