Zinc phenolsulfonate octahydrate, purified

Supplier: Spectrum Chemical Mfg. Corp.
Warning

Z1089-100GM Z1089-2.5KG Z1089-500GM Z1089-50KGBL
700016-334EA 148.6 USD
700016-334 700016-336 700016-338 700016-340
Zinc phenolsulfonate octahydrate, purified
Zinc phenolsulfonate octahydrate

Zinc Phenolsulfonate, Purified is used as an antiperspirant, an antiseptic, and in insecticide formulations.


The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Formula: Zn(C₆H₅O₄S)₂.8H₂O
Storage Temperature: Ambient
CAS Number: 1300-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR