Urease

Supplier: Spectrum Chemical Mfg. Corp.

UR115-078 UR115-885 UR115-U7 UR115-ZX
700011-512EA 2537.1 USD
700011-512 700011-514 700011-516 700011-518
Urease
Enzymes

Storage temperature - REFRIGERATE: Keep tightly closed.


GHS hazard : Health hazard

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR