L(-)-Tyrosine 98,5 USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

T1153-100GM T1153-1KG T1153-25GM T1153-50KGBL T1153-5KG T1153-25KGBL
95034-098EA 161.8 USD
95034-098 95034-100 95034-102 700007-482 700007-484 75812-546
L(-)-Tyrosine 98,5 USP
L(-)-Tyrosine

TYROSINE, USP is one of 22 acids that are used by cells in order to synthesize proteins. It is a non essential amino acid that has a polar side group. It is an important part of photosynthesis as it acts as an electron donor in the reduction of oxidized chlorophyll. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 285
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4
Merck Index: 13,09907

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 101.5 %
Specific Rotation [A]D -9.8 To-11.2
Loss On Drying max. 0.3 %
Related Compounds To Pass Test
Residue On Ignition max. 0.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR