Thiamine nitrate ≥98.0% USP
Supplier: Spectrum Chemical Mfg. Corp.
Thiamine Mononitrate, USP is the mononitrate salt form of a material also known as vitamin B1.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₂H₁₇N₅O₄S Storage Temperature: Ambient |
MDL Number:
MFCD00036330 CAS Number: 532-43-4 |
Specification Test Results
Assay (Dried Basis) | 98.0 - 102.0 % |
pH of a 2% Solution @ 25°C | 6.0 - 7.5 |
Loss On Drying | max. 1.0 % |
Residue On Ignition | max. 0.2 % |
Chloride (Cl) | max. 0.06 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...