Tamoxifen citrate ≥99.0% USP

Synonyms: (Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate

T1423-1KGBL
95036-186EA 16275.12 USD
95036-186
Tamoxifen citrate ≥99.0% USP
Tamoxifen citrate

Tamoxifen Citrate, USP is a chemotherapeutic agent used to treat breast cancer. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark;Health hazard

Formula: C₂₆H₂₉NO
MW: 563.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058321
CAS Number: 54965-24-1

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 101.0%
Loss On Drying max. 0.5%
Residue On Ignition max. 0.2%
E-Isomer max. 0.3%
Iron (Fe) max. 0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR