Sulfaguanidine

Supplier: Spectrum Chemical Mfg. Corp.

SU128-100GM SU128-500GM SU128-25GM
700009-284EA 105.85 USD
700009-284 700009-288 700009-286
Sulfaguanidine
Sulfaguanidine

Sulfaguanidine is used in veterinary medicine for the treatment of bacillary dysentery and other enteric infections.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₁₀N₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD00038136
CAS Number: 57-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR