Starch arrowroot, powder
Supplier: SPECTRUM CHEMICAL MFG CORP
S1551-2.5KG
S1551-500GM
700011-332EA
497.95
USD
700011-332
700011-334
Starch arrowroot, powder
Starch
Starch, Arrowroot, Powder is used in talcum powders and hair dyes. It is added to moisturizers as a thickening agent and to help active ingredients penetrate the upper levels of the skin. This product is used in cosmetics to help moisturizers penetrate the skin. It appears as a fine, white powder. It may have other uses similar to cornstarch. In cosmetics, it is used as an abrasive.
Formula:
(C₆H₁₀O₅)n MW: 162.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00082026 CAS Number: 9005-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...