Squalane ≥97.0% NF

Supplier: Spectrum Chemical Mfg. Corp.

S1987-100ML S1987-20LTBL S1987-500ML
95036-064EA 151.29 USD
95036-064 700002-310 95036-066
Squalane ≥97.0% NF
Squalane

Squalene, is an organic compound that be found occurring naturally in shark liver oil and certain plants. It is a hydrocarbon that all plants and animals produce in varying quantities. Mostly used in cosmetics as a natural skin moisturizer, it has recently been used as an immunologic adjuvant in certain vaccines.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₃₀H₆₂
Density: 0.812
Storage Temperature: Ambient
MDL Number: MFCD00008953
CAS Number: 111-01-3

Order Now

Specification Test Results

Assay 97.0 - 102.0 %
Specific Gravity @ 20°C 0.807 - 0.810
Refractive Index @ 20°C 1.4510 - 1.4525
Acid Value max. 0.2
Iodine Value max. 4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR