Oil orange SS

Supplier: Spectrum Chemical Mfg. Corp.

S1171-25GM S1171-1GM S1171-5GM
700010-330EA 879.15 USD
700010-330 700010-328 700010-332
Oil orange SS
Oil orange SS

Solvent Orange 2 is used as a dye for food, drugs, cosmetics, varnishes, oils, fats, and waxes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₇H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD00059523
CAS Number: 2646-17-5

Order Now

Specification Test Results

Absorption Max 505 NM
Melting Point 128.0 - 131.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR