Sodium sulfathiazole sesquihydrate ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sulphathiazole Sodium Salt, Sulfathiazole Sodium Salt

S1462-100GM S1462-500GM S1462-1KG
700015-740EA 231.35 USD
700015-740 700015-744 700015-742
Sodium sulfathiazole sesquihydrate ≥99%
Sodium sulfathiazole sesquihydrate

Sodium Sulfathiazole, Sesquihydrate, an organosulfur compound, can be used as a short-acting sulfa drug and as an oral and topical antimicrobial, Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₈N₃NaO₂S₂·1.5H₂O
MW: 277.3 g/mol (anhy)
Storage Temperature: Ambient
CAS Number: 144-74-1

Order Now

Specification Test Results

Assay 99 % min.
Loss On Drying max. 9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR