Sodium nitrate ≥99.0%, granulated FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

SO183-2.5KG SO183-12KG SO183-50KGBL SO183-500GM
75812-458EA 1548.59 USD
75812-458 95030-846 95030-844 700009-876
Sodium nitrate ≥99.0%, granulated FCC
Sodium nitrate

Sodium Nitrate, Granular, FCC is used as curing agent and food preservative that creates a distinct flavor, controls lipid oxidation, and acts as an antimicrobial. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame over circle;Exclamation mark
UN1498

Formula: NaNO₃
MW: 84.99 g/mol
Density: 2.260
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
UN: 1498
ADR: 5.1,III

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5 %
Lead (Pb) max. 4 mg/kg
Total Chlorine To Pass Test
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR