Silica gel Grade 60, 60A 60-200 µm

Supplier: SPECTRUM CHEMICAL MFG CORP

SIL57-25KGBL SIL57-100GM SIL57-1KG SIL57-5KG
700000-712EA 2954.28 USD
700000-712 700000-708 700000-710 700000-714
Silica gel Grade 60, 60A 60-200 µm
Silica gel

Silica Gel, Grade 60, 60-200 Micron, 60A is a form of silicon dioxide made from sodium silicate. It is granular and porous solid and is a naturally occurring mineral. It is often used as a desiccant to control humidity and avoid spoilage.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Specification Test Results

pH (Suspension 5% w/w) 6.0 - 8.0
Surface Area 480 - 550 M2/G
Under 45µm max. 15.0%
Over 200µm max. 10.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR