Sesame oil NF

Supplier: SPECTRUM CHEMICAL MFG CORP

SE130-20LT SE130-4CS4 SE130-4LTGL SE130-500CS6 SE130-20LTMT SE130-500MLG SE130-200LTB SE130-200LTB
89078-994EA 1735.25 USD
89078-994 95035-634 95035-636 95035-638 700000-146 95035-640 700000-144 89078-996
Sesame oil NF
Sesame oil

SESAME OIL, NF also known as til oil, is an oil derived from the sesame seed. As an edible vegetable oil, it is widely used in most Eastern and some Middle Eastern cuisine's as a flavor eNHancer. It is also used in the personal care industry in soap production, as cosmetic oil carrier and as the base for massage oils. Medicinally, its major use is as a solvent in the preparation of sustained-release intramuscular / subcutaneous injections of oil soluble actives such as steroids and avermectins. It is also used as an excipient in suspensions and emulsions, ointments, liniments, ophthalmic preparations, oral capsules and suppositories. Spectrum Chemical NF SESAME OIL products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers. Prevent exposure to excessive heat.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 


Caution: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Storage Temperature: Ambient
MDL Number: MFCD00132243
CAS Number: 8008-74-0

Order Now

Specification Test Results

Triglyceride Composition To Pass Test
Specific Gravity @ 25°C 0.912 - 0.921
Iodine Value 103 - 116
Saponification Value 188 - 195
Solidification Range of The Fatty Acids 20°- 25°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR