Safflower oil USP

Supplier: Spectrum Chemical Mfg. Corp.

S1004-200LTB S1004-4LTGL S1004-20LT S1004-500MLG
700011-906EA 3378.25 USD
700011-906 95035-598 700011-908 700011-910
Safflower oil USP
Safflower oil

SAFFLOWER OIL, USP is derived from the safflower plant. Safflower oil is used frequently to help treat fever, tumors, coughs, and breathing problems, along with clotting conditions, pain and traumatic injuries. It can also be used to induce sweating, as a laxative, and a stimulant. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Density: 0.921
Storage Temperature: Ambient
MDL Number: MFCD00132216
CAS Number: 8001-23-8

Order Now

Specification Test Results

Fatty Acid Composition To Pass Test
Alkaline Impurities To Pass Test
Free Fatty Acids To Pass Test
Peroxide max. 10.0Meq/KG
Unsaponifiable Matter max. 1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR