Rapeseed oil

Supplier: Spectrum Chemical Mfg. Corp.

R2010-500ML R2010-200LTB R2010-4LT
700000-194EA 114.51 USD
700000-194 700000-190 700000-192
Rapeseed oil
Rapeseed oil

Rapeseed Oil is used as a raw material in the production of biodiesel (rapeseed methyl ester).


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Refrigerator
MDL Number: MFCD00217791
CAS Number: 8002-13-9

Order Now

Specification Test Results

Erucic Acid (C22:1) 40 %
Specific Gravity 0.905 - 0.912
Iodine Value 97 - 115
Saponification Value 170 - 180
Free Fatty Acids (As Oleic Acid) max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR