Pyridoxine hydrochloride ≥98.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

PY103-100GM PY103-1KG PY103-25GM
700009-352EA 4474.24 USD
700009-352 95035-508 95035-510 95035-512 75812-106 75812-108
Pyridoxine hydrochloride ≥98.0% USP
Pyridoxine hydrochloride

Pyridoxine Hydrochloride, USP is a form of vitamin B6. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₈H₁₂ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00012807
CAS Number: 58-56-0

Order Now

Specification Test Results

Assay (C₈H₁₁NO₃[-HCl) 98.0 - 102.0 %
Chloride Content 16.9 - 17.6 %
Loss On Drying max. 0.5 %
Residue After Ignition max. 0.1 %
Heavy Metals max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR