Potassium palmitate 97

Supplier: SPECTRUM CHEMICAL MFG CORP

P1818-100GM P1818-25GM P1818-500GM
700011-856EA 321.55 USD
700011-856 700011-858 700011-860
Potassium palmitate 97
Potassium palmitate

Potassium Palmitate is used as a binder, emulsifier, and anti-caking agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00045930
CAS Number: 2624-31-9

Order Now

Specification Test Results

Assay Approx. 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR