Polyoxyethylene 7 coconut glycerides
Supplier: Spectrum Chemical Mfg. Corp.
Polyoxyethylene 7 Coconut Glycerides is used in personal care products for its moisturizing and emulsifying properties.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
CAS Number:
68553-03-7 |
Specification Test Results
Appearance @ 25° C | Liquid |
Saponification Value | 85 - 105 |
Color (Gardner) | max. 2 |
Acid Value | max. 1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...