Polyethyleneimine, M.W. ∼ 10,000 (by GPC), branched
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
Polyethylenimine, Branched, Av. Mn (by GPC) Approx. 10,000, also known as Polyaziridine, has many applications in products like cosmetics, detergents, and adhesives as a chelating agent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
(CH2CH2NH)n Density: 1.030 Storage Temperature: Ambient |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...