Polyethylene glycol monomethyl ether 2000

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH

P2070-1KG P2070-150LBB
700012-548EA 647.85 USD
700012-548 700012-550 700012-546
Polyethylene glycol monomethyl ether 2000
Polyethylene glycol monomethyl ether

Polyethylene Glycol 2000 Monomethyl Ether, commonly referred to as PEG 2000 MONOMETHYLETHER, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: CH₃(OCH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR