Polyacrylic acid sodium salt solution 45%

Supplier: Spectrum Chemical Mfg. Corp.

P1704-100GM P1704-500GM
700012-368EA 125.2 USD
700012-368 700012-370
Polyacrylic acid sodium salt solution 45%
Polyacrylic acid sodium salt

Polyacrylic Acid Sodium Salt, 45 Percent Solution, also known as PAAc or Acrysol, is an anionic polymer used in disposable diapers and as thickening and emulsifying agents in the pharmaceutical and cosmetic industries. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: [CH₂CH(CO₂Na)]n
Density: 1.23
Storage Temperature: Ambient
MDL Number: MFCD00147949
CAS Number: 9003-04-7

Order Now

Specification Test Results

Appearance Viscous Liquid
Viscosity 350 - 1000 cSt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR