Polyacrylic acid sodium salt
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Polyacrylic Acid Sodium Salt is used as a cosmetic binding, film forming and viscosity controlling agent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
[CH₂CH(CO₂Na)]n Storage Temperature: Ambient |
MDL Number:
MFCD00147949 CAS Number: 9003-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...