Sodium-4-hydroxybenzoate

Supplier: Spectrum Chemical Mfg. Corp.

H2211-100GM H2211-25GM H2211-25KGBL
700006-490EA 318.85 USD
700006-490 700006-492 700006-494
Sodium-4-hydroxybenzoate
Sodium-4-hydroxybenzoate

p-Hydroxybenzoic Acid Monosodium Salt is a chemical synthetic intermediate.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₆O₃
Storage Temperature: Ambient
MDL Number: MFCD00016530
CAS Number: 114-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR