Phosphatidylcholine
Supplier: Spectrum Chemical Mfg. Corp.
Phosphatidylcholine, also known as lecithin, has been shown to play a vital role in many important areas including maintaining cell structure, fat metabolism, memory, nerve signaling, as a precursor to important neurotransmitters, and liver health.
Storage Temperature:
Freezer |
CAS Number:
97281-47-5 |
Specification Test Results
Phosphatidyl Choline | 92 % min. |
Lyso-Phosphatidylcholine | 3 +/- 3 % |
(+/-)-Alpha-Tocopherol | 0.1 % min. |
Peroxide Value | max. 5 |
Acid Value | max. 0.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...