Petrolatum, yellow USP

Supplier: Spectrum Chemical Mfg. Corp.

PE130-2.5KG PE130-500GM PE130-15.9KG PE130-175KGB PE130-175KGB
95034-876EA 187.25 USD
95034-876 95034-878 700000-488 700000-490 89078-922
Petrolatum, yellow USP
Petrolatum

Petrolatum, Yellow, USP is a carrier or vehicle excipient. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD00147839
CAS Number: 8009-03-8

Order Now

Specification Test Results

Stabilizer (Butylhydroxytoluene) max. 0.002 %
Specific Gravity @60 C 0.815-0.880
Melting Range 38 - 60 °C
Color To Pass Test
Acidity To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR