Pectin (from apple), powder

Supplier: SPECTRUM CHEMICAL MFG CORP

A1201-12KG A1201-2.5KG A1201-500GM
700013-426EA 917.9 USD
700013-426 700013-428 700013-430
Pectin (from apple), powder
Pectin

Apple Pectin, Powder is used as a thickening agent in cosmetic preparations.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00081838
CAS Number: 9000-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR