Palmarosa oil FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

P1027-125ML P1027-500MLG P1027-500MLG
95034-824EA 244.2 USD
95034-824 700015-058 95034-826
Palmarosa oil FCC
Palmarosa oil

Palmarosa Oil, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Exclamation mark

Storage Temperature: Ambient
MDL Number: MFCD00241309
CAS Number: 8014-19-5

Order Now

Specification Test Results

Assay (Total Alcohols) 88.0 % min.
Assay (Esters As Geranyl Acetate) 4.0 - 18.0 %
Angular Rotation -2° to +3°
Refractive Index @ 20°C 1.470- 1.476
Solubility in Alcohol To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR