Sodium oxamate ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Oxamic acid sodium salt, Sodium 2-amino-2-oxoacetate

O2084-100GM O2084-25GM O2084-5GM
10860-338EA 956.55 USD
10860-338 10860-340 10860-342
Sodium oxamate ≥98%
Sodium oxamate
Formula: C₂H₂NNaO₃
MW: 111.03
Storage Temperature: Ambient
MDL Number: MFCD00044553
CAS Number: 565-73-1

Order Now

Specification Test Results

Assay Approx. 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR