N-Vinylformamide ≥96% stabilised

Supplier: SPECTRUM CHEMICAL MFG CORP


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

V1526-25GM V1526-500GM
10860-564EA 63.1 USD
10860-564 10860-566
N-Vinylformamide ≥96% stabilised
N-Vinylformamide

Stabilized with BHT Storage temperature - HEAT SENSITIVE: Refrigerate.


GHS hazard : Corrosion;Exclamation mark


Caution: Stabilized with BHT
Storage temperature - HEAT SENSITIVE: Refrigerate.

Formula: C₃H₅NO
MW: 71.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00081206
CAS Number: 13162-05-5

Order Now

Specification Test Results

Purity (GC) 96 %
Specific Gravity (20/20) 1.0190 - 1.0230
Refractive Index (N20/D) 1.4930 - 1.4970


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR