Nicotinic acid (Niacin) ≥99.5%, Powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 3-Pyridinecarboxylic acid, Pyridine-3-carboxylic acid, 3-Picolinic acid, Niacin, Nicotinic acid, Vitamin B3

N1013-1KG N1013-500GM N1013-100GM N1013-25KGBL
95034-724EA 331.3 USD
95034-724 95034-726 95034-722 75811-736
Nicotinic acid (Niacin) ≥99.5%, Powder FCC
Nicotinic acid (Niacin)

Niacin, Powder, FCC is a vitamin supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₆H₅NO₂
MW: 123.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6

Order Now

Specification Test Results

Assay (Dried Basis) 99.5-101.0 %
Melting Range 234 - 238 °C
Loss On Drying max. 1.0 %
Residue On Ignition max. 0.1 %
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR