Naproxen sodium ≥98.0% USP

Synonyms: Sodium (S)-2-(6-methoxynaphthalen-2-yl)propanoate

N1026-100GM N1026-25GM N1026-500GM N1026-1KGBL N1026-5GM N1026-12KGBL
95034-700EA 465.12 USD
95034-700 95034-698 75811-740 95034-704 75811-738 95034-702
Naproxen sodium ≥98.0% USP
Naproxen sodium

Naproxen Sodium, USP is a non-steroidal anti-inflammatory drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark;Health hazard

Formula: C₁₄H₁₃NaO₃
MW: 252.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058507
CAS Number: 26159-34-2

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0%
Specific Rotation -15.3° to -17.0
Chromatographic Purity To Pass Test
Loss On Drying max. 1.0%
Heavy Metals max. 0.002%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR