2-(Dimethylamino)-ethanol ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
2-Dimethylaminoethanol
, N,N-Dimethylethanolamine
, DMAE
D2399-2.5LTGL
D2399-500MLGL
D2399-20LT
10841-912EA
75.48
USD
10841-912
10841-910
10841-908
2-(Dimethylamino)-ethanol ≥99%
2-Dimethylaminoethanol
N,N-Dimethylethanolamine is a transparent, pale yellow liquid and primary alcohol that is used as a building block for the synthesis of cationic flocculants and ion exchange resins. It is also used used as a chemical intermediate for pharmaceuticals, dyes, corrosion inhibitors, and emulsifiers and as an additive to boiler water, paint removers, and amino resins.
GHS hazard : Corrosion;Flame;Skull and crossbones;Exclamation mark
UN2051
Formula:
C₄H₁₁NO MW: 89.14 g/mol Density: 0.887 Storage Temperature: Ambient |
MDL Number:
MFCD00002846 CAS Number: 108-01-0 UN: 2051 ADR: 8.3,II |
Specification Test Results
Assay | 99 % |
Color (APHA) | max. 20 |
Water | max. 0.1 % |
Identification | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...