Myrrh tincture ≥83%

Supplier: Spectrum Chemical Mfg. Corp.

MY115-100ML MY115-500MLG
700014-886EA 89.55 USD
700014-886 700014-888
Myrrh tincture ≥83%
Myrrh tincture

Myrrh Tincture, an aromatic resin from the tree species of the genus Commiphora, is used as an incense and in perfumes. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


GHS hazard : Flame;Health hazard;Exclamation mark
UN1170

Storage Temperature: Ambient
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

ALCH 83 - 88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR