Magnesium stearate BP, JP, NF
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Stearic acid Magnesium(II) salt, Magnesium(II) Stearate, Stearic acid magnesium salt
MA130-12KG
MA130-500CS6
MA130-2.5CS4
MA130-2.5KG
MA130-500GM
MA130-45KGBL
700005-416EA
1962.53
USD
700005-416
95034-260
95034-256
95034-258
95034-262
75811-702
Magnesium stearate BP, JP, NF
Magnesium stearate
Magnesium Stearate, NF, BP, JP is used as an anti-adherent in the manufacture of of medical capsules, tablets, and powders.
All Spectrum Chemical NF, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
C₃₆H₇₀MgO₄ MW: 591.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036391 CAS Number: 557-04-0 Merck Index: 13,05714 |
Specification Test Results
Assay (As MG; Dried Basis) | 4.0 - 5.0% |
Relative Content of Stearic & Palmitic Acid | To Pass Test |
Total Aerobic Microbial Count | max. 1000 Cfu/G |
Total Combined Molds & Yeasts Count | max. 500 Cfu/G |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...