Magnesium gluconate dihydrate ≥98.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

M1134-2.5KG M1134-100GM M1134-500GM
95034-208EA 1140.4 USD
95034-208 95034-206 95034-210
Magnesium gluconate dihydrate ≥98.0% FCC
Magnesium gluconate dihydrate

Magnesium Gluconate, Dihydrate, FCC is a mineral supplement used to prevent and treat low amounts of magnesium in the blood. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₂₂MgO₁₄·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150971
CAS Number: 59625-89-7

Order Now

Specification Test Results

Assay (Anhydrous Basis) 98.0-102.0 %
Water 3.0- 12.0 %
Chloride (Cl) max. 0.05 %
Lead (Pb) max. 2 mg/kg
Reducing Substances max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR