L-Theanine
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(S)-2-Amino-5-(ethylamino)-5-oxopentanoic acid, L-gamma-Glutamylethylamide, N-gamma-Ethyl-L-glutamine
T1298-5GM
T1298-1GM
700007-784EA
790.95
USD
700007-784
700007-782
L-Theanine
L-Theanine
L-Theanine is an amino acid and a glutamic acid derivative that is found in tea that is used as a nutritionial supplement.
Formula:
C₇H₁₄N₂O₃ MW: 174.20 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00059653 CAS Number: 3081-61-6 |
Specification Test Results
Appearance | White Crystallin |
Assay | Approx. 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...