Linalyl acetate
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Linalyl Acetate, also known as Bergamiol, is a phytochemical found in spice and flower plants which can be used as an adulterant in essential oils; it is also the acetate ester of linalool.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₂H₂₀O₂ Density: 0.895 Storage Temperature: Ambient |
MDL Number:
MFCD00008907 CAS Number: 115-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...