N,N-Dimethyl-1-dodecylamine N-oxide solution 29 - 31% in aqueous solution

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: N-ethyl-N-oxido-dodecan-1-amine, LDAO, Lauryldimethylamine oxide, N,N-Dimethyldodecan-1-amine oxide, Lauramine oxide, N,N-Dimethyl-N-dodecylamine N-oxide

L1014-5GM L1014-1GM
700006-792EA 881.85 USD
700006-792 700006-790
N,N-Dimethyl-1-dodecylamine N-oxide solution 29 - 31% in aqueous solution
N,N-Dimethyl-1-dodecylamine N-oxide

Lauramine Oxide, Solution is an amine oxide surfactant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


GHS hazard : Exclamation mark

Formula: C₁₄H₃₁NO
MW: 229.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002049
CAS Number: 1643-20-5

Order Now

Specification Test Results

Amine Oxide 29 - 31 %
pH (1 Part Solution Plus 9 Parts H₂O, at 25°C) 7 - 9
Free Peroxide max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR