L(+)-Asparagine monohydrate ≥98.0%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O
AS115-100GM
AS115-1KG
AS115-25GM
10860-576EA
138.69
USD
10860-576
10860-578
10860-580
L(+)-Asparagine monohydrate ≥98.0%
L(+)-Asparagine monohydrate
L-Asparagine, Monohydrate, also known as 2-Amino-3-carbamoylpropanoic acid, is one of the common amino acid on Earth. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
C₄H₈N₂O₃·1H₂O MW: 150.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151038 CAS Number: 5794-13-8 Merck Index: 14,00837 |
Specification Test Results
Assay (Dried Basis) | 98.0 - 101.5% |
Specific Rotation | +33.0° to +36.5 |
Loss On Drying | 11.5 - 12.5% |
Heavy Metals (As Pb) | max. 0.001% |
Lead (Pb) | max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...