Kinetin ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 6-(Furfurylamino)purine

K1139-100GM K1139-1KGBL K1139-25GM K1139-5GM
700007-032EA 2242.2 USD
700007-032 700007-036 700007-038 700007-040
Kinetin ≥99%
Kinetin

Kinetin is included in the plant hormone class that promotes cell division. It has the ability to induce cell division and has been used in plant tissue culture to induce formation of callus.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₉N₅O
Storage Temperature: Freezer
MDL Number: MFCD00075757
CAS Number: 525-79-1

Order Now

Specification Test Results

Assay 99% min.
Appearance White to Off-Whi
Loss On Drying max. 0.5%
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR